Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137167 MIM: 601468 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGCX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GGCX - gamma-glutamyl carboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000821.6 | Intron | NP_000812.2 | ||||
NM_001142269.3 | Intron | NP_001135741.1 | ||||
NM_001311312.1 | Intron | NP_001298241.1 | ||||
XM_005264259.4 | Intron | XP_005264316.1 | ||||
XM_011532764.2 | Intron | XP_011531066.1 | ||||
XM_011532765.2 | Intron | XP_011531067.1 | ||||
XM_017003803.1 | Intron | XP_016859292.1 |
MAT2A - methionine adenosyltransferase 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |