Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAAACCCTGTGGATTACGTGTAA[A/G]CTGAGTCATTCTTACAGTAAGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
60 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613952 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM46C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM46C - family with sequence similarity 46 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017709.3 | Intron | NP_060179.2 |
LOC100996263 - uncharacterized LOC100996263 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |