Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188855 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNLY PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GNLY - granulysin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302758.1 | 297 | Intron | NP_001289687.1 | |||
NM_006433.4 | 297 | Intron | NP_006424.2 | |||
NM_012483.3 | 297 | UTR 5 | NP_036615.2 | |||
XM_005264084.2 | 297 | Intron | XP_005264141.1 | |||
XM_005264085.2 | 297 | Intron | XP_005264142.1 | |||
XM_005264087.2 | 297 | UTR 5 | XP_005264144.1 |