Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188855 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNLY PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GNLY - granulysin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302758.1 | Intron | NP_001289687.1 | ||||
NM_006433.4 | Intron | NP_006424.2 | ||||
NM_012483.3 | Intron | NP_036615.2 | ||||
XM_005264084.2 | Intron | XP_005264141.1 | ||||
XM_005264085.2 | Intron | XP_005264142.1 | ||||
XM_005264087.2 | Intron | XP_005264144.1 |