Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCACCACATTACCGCATTTGGCT[C/T]CTAGCACGGCTCTATGAAATAATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600034 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EMX1 - empty spiracles homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004097.2 | 2046 | Intron | NP_004088.2 | |||
XM_005264203.3 | 2046 | Intron | XP_005264260.1 | |||
XM_011532697.2 | 2046 | UTR 5 | XP_011530999.1 |
Set Membership: |
HapMap |