Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609544 MIM: 605943 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCP110 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCP110 - centriolar coiled-coil protein 110 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199022.2 | Intron | NP_001185951.2 | ||||
NM_001323569.1 | Intron | NP_001310498.1 | ||||
NM_001323570.1 | Intron | NP_001310499.1 | ||||
NM_001323571.1 | Intron | NP_001310500.1 | ||||
NM_001323572.1 | Intron | NP_001310501.1 | ||||
NM_001323576.1 | Intron | NP_001310505.1 | ||||
NM_001323577.1 | Intron | NP_001310506.1 | ||||
NM_014711.5 | Intron | NP_055526.4 | ||||
XM_011545991.2 | Intron | XP_011544293.1 | ||||
XM_011545992.2 | Intron | XP_011544294.1 | ||||
XM_017023908.1 | Intron | XP_016879397.1 |
GDE1 - glycerophosphodiester phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |