Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFT2D3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SFT2D3 - SFT2 domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032740.3 | 2360 | UTR 3 | NP_116129.3 |
WDR33 - WD repeat domain 33 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006622.2 | 2360 | Intron | NP_001006623.1 | |||
NM_001006623.2 | 2360 | Intron | NP_001006624.1 | |||
NM_018383.4 | 2360 | Intron | NP_060853.3 | |||
XM_005263697.2 | 2360 | Intron | XP_005263754.1 | |||
XM_011511436.1 | 2360 | Intron | XP_011509738.1 | |||
XM_017004435.1 | 2360 | Intron | XP_016859924.1 | |||
XM_017004436.1 | 2360 | Intron | XP_016859925.1 |
Set Membership: |
HapMap |