Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAACCCACATTCTCCAAAGAAACA[C/T]GCCCTGCCTTGTTTACCCCCAGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605771 MIM: 607442 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX7A2L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
COX7A2L - cytochrome c oxidase subunit 7A2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319036.1 | Intron | NP_001305965.1 | ||||
NM_001319037.1 | Intron | NP_001305966.1 | ||||
NM_001319038.1 | Intron | NP_001305967.1 | ||||
NM_001319040.1 | Intron | NP_001305969.1 | ||||
NM_004718.3 | Intron | NP_004709.2 |
EML4 - echinoderm microtubule associated protein like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |