Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCTAGCTCTGTCTGCTAAAAGG[A/G]CCAAAAGGGAGACACCACAGTAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603501 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARG PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PARG - poly(ADP-ribose) glycohydrolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303486.1 | Intron | NP_001290415.1 | ||||
NM_001303487.1 | Intron | NP_001290416.1 | ||||
NM_001303489.1 | Intron | NP_001290418.1 | ||||
NM_001324381.1 | Intron | NP_001311310.1 | ||||
NM_003631.3 | Intron | NP_003622.2 | ||||
XM_011540305.1 | Intron | XP_011538607.1 | ||||
XM_011540308.2 | Intron | XP_011538610.1 |
Set Membership: |
HapMap |