Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGTTTTAGATTTTCTTCAGATTC[G/T]AAACTTCTTAGTTTTGCTTTCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601896 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TRAF3 - TNF receptor associated factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199427.1 | Intron | NP_001186356.1 | ||||
NM_003300.3 | Intron | NP_003291.2 | ||||
NM_145725.2 | Intron | NP_663777.1 | ||||
NM_145726.2 | Intron | NP_663778.1 | ||||
XM_011537116.2 | Intron | XP_011535418.1 | ||||
XM_011537117.2 | Intron | XP_011535419.1 | ||||
XM_011537118.2 | Intron | XP_011535420.1 | ||||
XM_017021617.1 | Intron | XP_016877106.1 | ||||
XM_017021618.1 | Intron | XP_016877107.1 | ||||
XM_017021619.1 | Intron | XP_016877108.1 | ||||
XM_017021620.1 | Intron | XP_016877109.1 |
Set Membership: |
Validated |