Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGCCCTTCTGAACACTGTTATC[C/T]GCAACACAGTTACCTGTTTATGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604763 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGEF12 - Rho guanine nucleotide exchange factor 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198665.1 | Intron | NP_001185594.1 | ||||
NM_001301084.1 | Intron | NP_001288013.1 | ||||
NM_015313.2 | Intron | NP_056128.1 | ||||
XM_006718805.3 | Intron | XP_006718868.1 | ||||
XM_011542720.2 | Intron | XP_011541022.1 | ||||
XM_017017420.1 | Intron | XP_016872909.1 | ||||
XM_017017421.1 | Intron | XP_016872910.1 | ||||
XM_017017422.1 | Intron | XP_016872911.1 |
Set Membership: |
HapMap |