Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTAGTTAGGTGTCAACATTATAG[C/T]TCTTCTGAAGATGTAAGAATTTTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604888 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GTF3C3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GTF3C3 - general transcription factor IIIC subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206774.1 | Intron | NP_001193703.1 | ||||
NM_012086.4 | Intron | NP_036218.1 | ||||
XM_005246965.4 | Intron | XP_005247022.1 |
Set Membership: |
HapMap |