Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAACTAAAGCTGGCAACTGGGA[A/C]ATCGTCCGGTGCTGCAGGTCTCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604508 MIM: 137028 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
COPS2 - COP9 signalosome subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143887.1 | 39 | Intron | NP_001137359.1 | |||
NM_004236.3 | 39 | Intron | NP_004227.1 |
GALK2 - galactokinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001556.2 | 39 | UTR 5 | NP_001001556.1 | |||
NM_001289030.1 | 39 | Intron | NP_001275959.1 | |||
NM_001289031.1 | 39 | Intron | NP_001275960.1 | |||
NM_002044.3 | 39 | Intron | NP_002035.1 | |||
XM_005254279.3 | 39 | Intron | XP_005254336.1 | |||
XM_005254280.3 | 39 | UTR 5 | XP_005254337.1 | |||
XM_005254284.3 | 39 | UTR 5 | XP_005254341.1 | |||
XM_006720461.3 | 39 | Intron | XP_006720524.1 | |||
XM_006720462.3 | 39 | Intron | XP_006720525.1 | |||
XM_006720463.3 | 39 | Intron | XP_006720526.1 | |||
XM_006720464.3 | 39 | Intron | XP_006720527.1 | |||
XM_011521441.2 | 39 | Intron | XP_011519743.1 | |||
XM_017022062.1 | 39 | Intron | XP_016877551.1 | |||
XM_017022063.1 | 39 | UTR 5 | XP_016877552.1 | |||
XM_017022064.1 | 39 | Intron | XP_016877553.1 | |||
XM_017022065.1 | 39 | UTR 5 | XP_016877554.1 | |||
XM_017022066.1 | 39 | Intron | XP_016877555.1 |
NDUFAF4P1 - NADH:ubiquinone oxidoreductase complex assembly factor 4 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |