Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604024 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC5A6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SLC5A6 - solute carrier family 5 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021095.2 | 1732 | Missense Mutation | TCC,TTC | S,F 481 | NP_066918.2 | |
XM_006712128.1 | 1732 | Missense Mutation | TCC,TTC | S,F 481 | XP_006712191.1 | |
XM_006712129.1 | 1732 | Missense Mutation | TCC,TTC | S,F 481 | XP_006712192.1 | |
XM_006712130.1 | 1732 | Missense Mutation | TCC,TTC | S,F 481 | XP_006712193.1 | |
XM_011533146.2 | 1732 | Missense Mutation | TCC,TTC | S,F 258 | XP_011531448.1 | |
XM_017005216.1 | 1732 | Missense Mutation | TCC,TTC | S,F 272 | XP_016860705.1 |
Set Membership: |
HapMap DME Validated Inventoried |