Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603582 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPM1F PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PPM1F - protein phosphatase, Mg2+/Mn2+ dependent 1F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOP3B - topoisomerase (DNA) III beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282112.1 | Intron | NP_001269041.1 | ||||
NM_001282113.1 | Intron | NP_001269042.1 | ||||
NM_003935.4 | Intron | NP_003926.1 | ||||
XM_005261811.1 | Intron | XP_005261868.1 | ||||
XM_005261813.2 | Intron | XP_005261870.1 | ||||
XM_006724349.1 | Intron | XP_006724412.1 | ||||
XM_006724350.1 | Intron | XP_006724413.1 | ||||
XM_011530482.1 | Intron | XP_011528784.1 | ||||
XM_017029038.1 | Intron | XP_016884527.1 | ||||
XM_017029039.1 | Intron | XP_016884528.1 | ||||
XM_017029040.1 | Intron | XP_016884529.1 | ||||
XM_017029041.1 | Intron | XP_016884530.1 |
Set Membership: |
HapMap Validated |