Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGCGGGGAGGTGCCAGAAAGAC[C/T]TGAGCAACCCGCTGGGCCCCTCTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614502 MIM: 611971 | ||||||||||||||||||||
Literature Links: |
C9orf116 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C9orf116 - chromosome 9 open reading frame 116 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101928525 - uncharacterized LOC101928525 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPS2 - mitochondrial ribosomal protein S2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016034.4 | Intron | NP_057118.1 | ||||
XM_006717136.3 | Intron | XP_006717199.1 |