Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGGTAGCAGTGAGGATGGCACA[A/C]ATTTATGTAGCCAGGCCACTCCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604872 MIM: 600818 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100652768 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100652768 - uncharacterized LOC100652768 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCSK7 - proprotein convertase subtilisin/kexin type 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004716.3 | Intron | NP_004707.2 |
SIDT2 - SID1 transmembrane family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAGLN - transgelin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001522.1 | Intron | NP_001001522.1 | ||||
NM_003186.3 | Intron | NP_003177.2 |