Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCAGTTGTTCTGTATACTTAAA[A/C]TGCTACTTTGCAGTACTCTGAGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM76B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM76B - family with sequence similarity 76 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144664.4 | Intron | NP_653265.3 | ||||
XM_005273775.4 | Intron | XP_005273832.1 | ||||
XM_005273776.4 | Intron | XP_005273833.1 | ||||
XM_011542610.2 | Intron | XP_011540912.1 | ||||
XM_011542611.2 | Intron | XP_011540913.1 | ||||
XM_011542612.1 | Intron | XP_011540914.1 | ||||
XM_011542613.1 | Intron | XP_011540915.1 |