Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609233 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDT13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUDT13 - nudix hydrolase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283014.1 | Intron | NP_001269943.1 | ||||
NM_001283015.1 | Intron | NP_001269944.1 | ||||
NM_001283016.1 | Intron | NP_001269945.1 | ||||
NM_001283017.1 | Intron | NP_001269946.1 | ||||
NM_001283019.1 | Intron | NP_001269948.1 | ||||
NM_015901.5 | Intron | NP_056985.3 | ||||
XM_017016033.1 | Intron | XP_016871522.1 |
Set Membership: |
HapMap |