Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCTGAGGCACGAGAATCACATGA[G/T]CTTGGAAAGTGGAGGTTTCAGTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601263 | ||||||||||||||||||||
Literature Links: |
MAP2K2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP2K2 - mitogen-activated protein kinase kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030662.3 | Intron | NP_109587.1 | ||||
XM_006722799.2 | Intron | XP_006722862.1 | ||||
XM_017026989.1 | Intron | XP_016882478.1 | ||||
XM_017026990.1 | Intron | XP_016882479.1 | ||||
XM_017026991.1 | Intron | XP_016882480.1 |