Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607099 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2CD2L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C2CD2L - C2CD2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HINFP - histone H4 transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243259.1 | Intron | NP_001230188.1 | ||||
NM_015517.4 | Intron | NP_056332.2 | ||||
NM_198971.2 | Intron | NP_945322.1 | ||||
XM_011542744.2 | Intron | XP_011541046.1 | ||||
XM_011542745.2 | Intron | XP_011541047.1 | ||||
XM_011542746.1 | Intron | XP_011541048.1 | ||||
XM_017017499.1 | Intron | XP_016872988.1 | ||||
XM_017017500.1 | Intron | XP_016872989.1 | ||||
XM_017017501.1 | Intron | XP_016872990.1 | ||||
XM_017017502.1 | Intron | XP_016872991.1 | ||||
XM_017017503.1 | Intron | XP_016872992.1 | ||||
XM_017017504.1 | Intron | XP_016872993.1 | ||||
XM_017017505.1 | Intron | XP_016872994.1 | ||||
XM_017017506.1 | Intron | XP_016872995.1 | ||||
XM_017017507.1 | Intron | XP_016872996.1 | ||||
XM_017017508.1 | Intron | XP_016872997.1 | ||||
XM_017017509.1 | Intron | XP_016872998.1 |
Set Membership: |
HapMap |