Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608889 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACMSD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACMSD - aminocarboxymuconate semialdehyde decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307983.1 | Intron | NP_001294912.1 | ||||
NM_138326.2 | Intron | NP_612199.2 | ||||
XM_005263586.4 | Intron | XP_005263643.1 | ||||
XM_005263588.4 | Intron | XP_005263645.1 | ||||
XM_005263589.4 | Intron | XP_005263646.1 | ||||
XM_005263590.4 | Intron | XP_005263647.1 | ||||
XM_011510592.2 | Intron | XP_011508894.1 | ||||
XM_017003325.1 | Intron | XP_016858814.1 | ||||
XM_017003326.1 | Intron | XP_016858815.1 |
Set Membership: |
HapMap |