Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGTAGCTTTACAGTGGAGAAATA[A/T]GTCAACAGACCACCTTAGCCAAGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613760 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC36A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC36A4 - solute carrier family 36 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286139.1 | Intron | NP_001273068.1 | ||||
NM_152313.3 | Intron | NP_689526.2 | ||||
XM_017017174.1 | Intron | XP_016872663.1 | ||||
XM_017017175.1 | Intron | XP_016872664.1 | ||||
XM_017017176.1 | Intron | XP_016872665.1 | ||||
XM_017017177.1 | Intron | XP_016872666.1 |
Set Membership: |
HapMap |