Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602422 MIM: 605303 MIM: 615975 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLBP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLBP - stem-loop binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TACC3 - transforming acidic coiled-coil containing protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006342.2 | Intron | NP_006333.1 | ||||
XM_005247929.2 | Intron | XP_005247986.1 | ||||
XM_005247930.2 | Intron | XP_005247987.1 | ||||
XM_011513386.2 | Intron | XP_011511688.1 | ||||
XM_017007653.1 | Intron | XP_016863142.1 |
TMEM129 - transmembrane protein 129 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |