Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142987 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAGLR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAGLR - HOXD antisense growth-associated long non-coding RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXD1 - homeobox D1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024501.2 | 1150 | Missense Mutation | ACC,GCC | T,A 296 | NP_078777.1 | |
XM_005246507.3 | 1150 | UTR 3 | XP_005246564.1 |
MIR7704 - microRNA 7704 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |