Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCTGATGGTGCCTTTCTTTCTG[G/T]GCTTTCTTTTCTCTTGCACTGACTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614248 | ||||||||||||||||||||
Literature Links: |
LINC00597 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LINC00597 - long intergenic non-protein coding RNA 597 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PEAK1 - pseudopodium enriched atypical kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024776.3 | Intron | NP_079052.2 | ||||
XM_005254670.2 | Intron | XP_005254727.1 | ||||
XM_005254671.2 | Intron | XP_005254728.1 | ||||
XM_005254673.2 | Intron | XP_005254730.1 | ||||
XM_005254674.2 | Intron | XP_005254731.1 | ||||
XM_005254675.4 | Intron | XP_005254732.1 | ||||
XM_011522033.2 | Intron | XP_011520335.1 | ||||
XM_011522034.2 | Intron | XP_011520336.1 | ||||
XM_011522035.2 | Intron | XP_011520337.1 | ||||
XM_011522036.2 | Intron | XP_011520338.1 | ||||
XM_011522037.2 | Intron | XP_011520339.1 | ||||
XM_011522038.2 | Intron | XP_011520340.1 | ||||
XM_011522039.2 | Intron | XP_011520341.1 | ||||
XM_011522040.1 | Intron | XP_011520342.1 | ||||
XM_011522041.1 | Intron | XP_011520343.1 | ||||
XM_011522042.2 | Intron | XP_011520344.1 |