Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTCTGTGGCAGCCCCCACTCCC[A/C]ACCCCGCCCAGCCCTGCCGGCCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605719 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAT2 - linker for activation of T-cells family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014146.3 | Intron | NP_054865.2 | ||||
NM_032463.2 | Intron | NP_115852.1 | ||||
NM_032464.2 | Intron | NP_115853.2 | ||||
XM_011516558.2 | Intron | XP_011514860.1 |