Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107680 MIM: 107690 MIM: 107720 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
APOA1 - apolipoprotein A1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOA1-AS - APOA1 antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOA4 - apolipoprotein A4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOC3 - apolipoprotein C3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000040.1 | Intron | NP_000031.1 |
Set Membership: |
Validated |