Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611636 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAALAD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NAALAD2 - N-acetylated alpha-linked acidic dipeptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300930.1 | 55 | UTR 5 | NP_001287859.1 | |||
NM_005467.3 | 55 | UTR 5 | NP_005458.1 | |||
XM_017017043.1 | 55 | Intron | XP_016872532.1 | |||
XM_017017044.1 | 55 | Intron | XP_016872533.1 | |||
XM_017017045.1 | 55 | UTR 5 | XP_016872534.1 | |||
XM_017017046.1 | 55 | Intron | XP_016872535.1 |
Set Membership: |
HapMap |