Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
52 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601365 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DVL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
DVL1 - dishevelled segment polarity protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MXRA8 - matrix remodeling associated 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282582.1 | 1780 | UTR 3 | NP_001269511.1 | |||
NM_001282583.1 | 1780 | UTR 3 | NP_001269512.1 | |||
NM_001282584.1 | 1780 | UTR 3 | NP_001269513.1 | |||
NM_001282585.1 | 1780 | Intron | NP_001269514.1 | |||
NM_032348.3 | 1780 | UTR 3 | NP_115724.1 | |||
XM_017001516.1 | 1780 | Intron | XP_016857005.1 | |||
XM_017001517.1 | 1780 | Intron | XP_016857006.1 |