Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGCCACTTAGACCACAGATTCC[A/G]AGCAGGCTCATTGGCTTTTCCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612314 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GSTO2 - glutathione S-transferase omega 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITPRIP - inositol 1,4,5-trisphosphate receptor interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272012.1 | 4950 | UTR 3 | NP_001258941.1 | |||
NM_001272013.1 | 4950 | UTR 3 | NP_001258942.1 | |||
NM_033397.3 | 4950 | UTR 3 | NP_203755.1 | |||
XM_005270257.2 | 4950 | UTR 3 | XP_005270314.1 |