Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612314 | ||||||||||||||||||||||||||||||||
Literature Links: |
GSTO2 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
GSTO2 - glutathione S-transferase omega 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ITPRIP - inositol 1,4,5-trisphosphate receptor interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272012.1 | 3424 | UTR 3 | NP_001258941.1 | |||
NM_001272013.1 | 3424 | UTR 3 | NP_001258942.1 | |||
NM_033397.3 | 3424 | UTR 3 | NP_203755.1 | |||
XM_005270257.2 | 3424 | UTR 3 | XP_005270314.1 |