Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATCAAGATGTAGGCTGTTGACCT[A/G]TATACTTACAACTAATATTGCAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607124 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLPP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLPP1 - phospholipid phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003711.3 | Intron | NP_003702.2 | ||||
NM_176895.2 | Intron | NP_795714.1 | ||||
XM_006714724.2 | Intron | XP_006714787.1 |
SKIV2L2 - Ski2 like RNA helicase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015360.4 | Intron | NP_056175.3 |