Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCACACAGGGGAAACAACTTTTC[A/C]TTATGAAAAACTGGGAGACTGACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600909 MIM: 603294 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LSS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LSS - lanosterol synthase (2,3-oxidosqualene-lanosterol cyclase) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCM3AP - minichromosome maintenance complex component 3 associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003906.4 | Intron | NP_003897.2 | ||||
XM_005261203.4 | Intron | XP_005261260.1 | ||||
XM_005261204.4 | Intron | XP_005261261.1 | ||||
XM_005261205.3 | Intron | XP_005261262.1 |
MCM3AP-AS1 - MCM3AP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |