Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRDM15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PRDM15 - PR domain 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040424.2 | 5380 | UTR 3 | NP_001035514.1 | |||
NM_001282934.1 | 5380 | UTR 3 | NP_001269863.1 | |||
NM_022115.4 | 5380 | Intron | NP_071398.3 | |||
XM_006724039.2 | 5380 | UTR 3 | XP_006724102.1 | |||
XM_006724040.2 | 5380 | UTR 3 | XP_006724103.1 | |||
XM_011529674.1 | 5380 | UTR 3 | XP_011527976.1 | |||
XM_011529675.1 | 5380 | UTR 3 | XP_011527977.1 | |||
XM_011529676.2 | 5380 | UTR 3 | XP_011527978.1 | |||
XM_011529677.2 | 5380 | UTR 3 | XP_011527979.1 | |||
XM_011529678.2 | 5380 | UTR 3 | XP_011527980.1 | |||
XM_011529679.2 | 5380 | UTR 3 | XP_011527981.1 | |||
XM_011529681.2 | 5380 | UTR 3 | XP_011527983.1 | |||
XM_011529683.1 | 5380 | UTR 3 | XP_011527985.1 | |||
XM_017028425.1 | 5380 | UTR 3 | XP_016883914.1 | |||
XM_017028426.1 | 5380 | UTR 3 | XP_016883915.1 |
Set Membership: |
HapMap |