Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAATGATGTGAGTAAATTCTTCA[C/T]GTTTTTTTCTTTAAACTGGGAAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603152 MIM: 600609 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5J PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ATP5J - ATP synthase, H+ transporting, mitochondrial Fo complex subunit F6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003696.1 | Intron | NP_001003696.1 | ||||
NM_001003697.1 | Intron | NP_001003697.1 | ||||
NM_001003701.1 | Intron | NP_001003701.1 | ||||
NM_001003703.1 | Intron | NP_001003703.1 | ||||
NM_001320266.1 | Intron | NP_001307195.1 | ||||
NM_001320267.1 | Intron | NP_001307196.1 | ||||
NM_001685.4 | Intron | NP_001676.2 |
GABPA - GA binding protein transcription factor alpha subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
Validated |