Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAAATTTCGACGGGGTCTTCTCAC[C/G]GGTTTCATTCAGTTGGCCACTGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602436 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC102725068 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC102725068 - uncharacterized LOC102725068 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MICB - MHC class I polypeptide-related sequence B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289160.1 | 89 | Intron | NP_001276089.1 | |||
NM_001289161.1 | 89 | UTR 5 | NP_001276090.1 | |||
NM_005931.4 | 89 | UTR 5 | NP_005922.2 |
Set Membership: |
HapMap Validated |