Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601833 MIM: 142580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIF1 - allograft inflammatory factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6832 - microRNA 6832 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRRC2A - proline rich coiled-coil 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004638.3 | Intron | NP_004629.3 | ||||
NM_080686.2 | Intron | NP_542417.2 | ||||
XM_017011274.1 | Intron | XP_016866763.1 |
SNORA38 - small nucleolar RNA, H/ACA box 38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |