Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604700 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TOM1 - target of myb1 membrane trafficking protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135729.1 | Intron | NP_001129201.1 | ||||
NM_001135730.1 | Intron | NP_001129202.1 | ||||
NM_001135732.1 | Intron | NP_001129204.1 | ||||
NM_005488.2 | Intron | NP_005479.1 | ||||
XM_011529818.2 | Intron | XP_011528120.1 | ||||
XM_011529820.2 | Intron | XP_011528122.1 | ||||
XM_017028529.1 | Intron | XP_016884018.1 | ||||
XM_017028530.1 | Intron | XP_016884019.1 |