Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116790 MIM: 606448 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
COMT - catechol-O-methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000754.3 | Intron | NP_000745.1 | ||||
NM_001135161.1 | Intron | NP_001128633.1 | ||||
NM_001135162.1 | Intron | NP_001128634.1 | ||||
NM_007310.2 | Intron | NP_009294.1 | ||||
XM_011529886.1 | Intron | XP_011528188.1 | ||||
XM_017028594.1 | Intron | XP_016884083.1 | ||||
XM_017028595.1 | Intron | XP_016884084.1 |
TXNRD2 - thioredoxin reductase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |