Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATAAGGAAGGCTGACCAGATTCTC[C/T]GATGAGACATAGTGATGATATCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613840 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF304 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF304 - zinc finger protein 304 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290318.1 | Intron | NP_001277247.1 | ||||
NM_001290319.1 | Intron | NP_001277248.1 | ||||
NM_020657.3 | Intron | NP_065708.2 | ||||
XM_011527145.2 | Intron | XP_011525447.1 | ||||
XM_017027024.1 | Intron | XP_016882513.1 |
Set Membership: |
HapMap |