Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603481 MIM: 604481 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMD13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PSMD13 - proteasome 26S subunit, non-ATPase 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002817.3 | 413 | Missense Mutation | AAC,AGC | N,S 13 | NP_002808.3 | |
NM_175932.2 | 413 | Missense Mutation | AAC,AGC | N,S 13 | NP_787128.2 | |
XM_011520235.2 | 413 | Missense Mutation | AAC,AGC | N,S 13 | XP_011518537.1 |
SIRT3 - sirtuin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017524.2 | 413 | Intron | NP_001017524.1 | |||
NM_012239.5 | 413 | Intron | NP_036371.1 | |||
XM_005252835.1 | 413 | Intron | XP_005252892.1 | |||
XM_011519956.1 | 413 | Intron | XP_011518258.1 | |||
XM_011519957.1 | 413 | Intron | XP_011518259.1 | |||
XM_017017428.1 | 413 | Intron | XP_016872917.1 | |||
XM_017017429.1 | 413 | Intron | XP_016872918.1 | |||
XM_017017430.1 | 413 | Intron | XP_016872919.1 | |||
XM_017017431.1 | 413 | Intron | XP_016872920.1 |
Set Membership: |
HapMap |