Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGGTACTGTTTCAGGTACATAA[C/T]GGCTTTGGTTTAACATGAAAGCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300097 MIM: 300153 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAGEB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAGEB1 - MAGE family member B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002363.4 | Intron | NP_002354.2 | ||||
NM_177404.2 | Intron | NP_796379.1 | ||||
NM_177415.2 | Intron | NP_803134.1 |
MAGEB4 - MAGE family member B4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |