Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606184 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADAMTS12 - ADAM metallopeptidase with thrombospondin type 1 motif 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324511.1 | 6072 | Intron | NP_001311440.1 | |||
NM_001324512.1 | 6072 | UTR 3 | NP_001311441.1 | |||
NM_030955.3 | 6072 | UTR 3 | NP_112217.2 | |||
XM_017009905.1 | 6072 | UTR 3 | XP_016865394.1 | |||
XM_017009906.1 | 6072 | UTR 3 | XP_016865395.1 | |||
XM_017009907.1 | 6072 | UTR 3 | XP_016865396.1 | |||
XM_017009908.1 | 6072 | UTR 3 | XP_016865397.1 | |||
XM_017009909.1 | 6072 | UTR 3 | XP_016865398.1 |
Set Membership: |
HapMap Validated |