Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164005 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFIX PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NFIX - nuclear factor I X | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271043.2 | Intron | NP_001257972.1 | ||||
NM_001271044.2 | Intron | NP_001257973.1 | ||||
NM_002501.3 | Intron | NP_002492.2 | ||||
XM_005259917.4 | Intron | XP_005259974.1 | ||||
XM_005259918.4 | Intron | XP_005259975.1 | ||||
XM_005259919.3 | Intron | XP_005259976.1 | ||||
XM_005259920.3 | Intron | XP_005259977.1 | ||||
XM_006722760.2 | Intron | XP_006722823.1 | ||||
XM_011528040.1 | Intron | XP_011526342.1 | ||||
XM_017026837.1 | Intron | XP_016882326.1 |
Set Membership: |
HapMap |