Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCAGACAGCTCTTTTATTGACTT[C/T]GTCTGCTTGGTGCGGGGGTTGGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611621 MIM: 601313 MIM: 191092 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371049 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371049 - uncharacterized LOC105371049 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1225 - microRNA 1225 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PKD1 - polycystin 1, transient receptor potential channel interacting | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000296.3 | 14136 | UTR 3 | NP_000287.3 | |||
NM_001009944.2 | 14136 | UTR 3 | NP_001009944.2 | |||
XM_005255370.3 | 14136 | UTR 3 | XP_005255427.1 | |||
XM_011522528.2 | 14136 | UTR 3 | XP_011520830.1 | |||
XM_011522529.2 | 14136 | UTR 3 | XP_011520831.1 | |||
XM_011522530.2 | 14136 | UTR 3 | XP_011520832.1 | |||
XM_011522532.2 | 14136 | UTR 3 | XP_011520834.1 | |||
XM_011522534.2 | 14136 | UTR 3 | XP_011520836.1 | |||
XM_011522535.2 | 14136 | UTR 3 | XP_011520837.1 | |||
XM_011522537.2 | 14136 | UTR 3 | XP_011520839.1 | |||
XM_017023285.1 | 14136 | UTR 3 | XP_016878774.1 |
TSC2 - tuberous sclerosis 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000548.4 | 14136 | Intron | NP_000539.2 | |||
NM_001077183.2 | 14136 | Intron | NP_001070651.1 | |||
NM_001114382.2 | 14136 | Intron | NP_001107854.1 | |||
NM_001318827.1 | 14136 | Intron | NP_001305756.1 | |||
NM_001318829.1 | 14136 | Intron | NP_001305758.1 | |||
NM_001318831.1 | 14136 | Intron | NP_001305760.1 | |||
NM_001318832.1 | 14136 | Intron | NP_001305761.1 | |||
XM_005255529.4 | 14136 | Intron | XP_005255586.2 | |||
XM_005255531.4 | 14136 | Intron | XP_005255588.2 | |||
XM_011522636.2 | 14136 | Intron | XP_011520938.1 | |||
XM_011522637.2 | 14136 | Intron | XP_011520939.1 | |||
XM_011522638.2 | 14136 | Intron | XP_011520940.2 | |||
XM_011522639.2 | 14136 | Intron | XP_011520941.1 | |||
XM_011522640.2 | 14136 | Intron | XP_011520942.1 | |||
XM_017023615.1 | 14136 | Intron | XP_016879104.1 | |||
XM_017023616.1 | 14136 | Intron | XP_016879105.1 | |||
XM_017023617.1 | 14136 | Intron | XP_016879106.1 | |||
XM_017023618.1 | 14136 | Intron | XP_016879107.1 |