Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613202 MIM: 602428 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHTF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CHTF8 - chromosome transmission fidelity factor 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HAS3 - hyaluronan synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199280.1 | 370 | Silent Mutation | GCA,GCG | A,A 93 | NP_001186209.1 | |
NM_005329.2 | 370 | Silent Mutation | GCA,GCG | A,A 93 | NP_005320.2 | |
NM_138612.2 | 370 | Silent Mutation | GCA,GCG | A,A 93 | NP_619515.1 | |
XM_005255921.2 | 370 | Silent Mutation | GCA,GCG | A,A 93 | XP_005255978.1 | |
XM_011523061.2 | 370 | Silent Mutation | GCA,GCG | A,A 93 | XP_011521363.1 |
Set Membership: |
HapMap Validated |