Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACATAGGTCACAGAATTCTCAC[A/C]ATTCACCTGACCCTTTGAAGACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604056 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS3ST2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HS3ST2 - heparan sulfate-glucosamine 3-sulfotransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006043.1 | Intron | NP_006034.1 | ||||
XM_011546001.2 | Intron | XP_011544303.1 | ||||
XM_011546002.2 | Intron | XP_011544304.1 |
LOC107984851 - collagen alpha-1(XVI) chain-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |