Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600608 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
P4HA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
P4HA2 - prolyl 4-hydroxylase subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017973.1 | Intron | NP_001017973.1 | ||||
NM_001017974.1 | Intron | NP_001017974.1 | ||||
NM_001142598.1 | Intron | NP_001136070.1 | ||||
NM_001142599.1 | Intron | NP_001136071.1 | ||||
NM_004199.2 | Intron | NP_004190.1 | ||||
XM_005272116.4 | Intron | XP_005272173.1 | ||||
XM_005272117.4 | Intron | XP_005272174.1 | ||||
XM_005272118.4 | Intron | XP_005272175.1 | ||||
XM_005272119.4 | Intron | XP_005272176.1 | ||||
XM_005272120.4 | Intron | XP_005272177.1 | ||||
XM_006714728.3 | Intron | XP_006714791.1 | ||||
XM_006714729.3 | Intron | XP_006714792.1 | ||||
XM_006714730.3 | Intron | XP_006714793.1 | ||||
XM_011543705.2 | Intron | XP_011542007.1 | ||||
XM_017010011.1 | Intron | XP_016865500.1 | ||||
XM_017010012.1 | Intron | XP_016865501.1 |
P4HA2-AS1 - P4HA2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |