Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164920 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KIT - KIT proto-oncogene receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000222.2 | Intron | NP_000213.1 | ||||
NM_001093772.1 | Intron | NP_001087241.1 | ||||
XM_005265740.1 | Intron | XP_005265797.1 | ||||
XM_005265741.1 | Intron | XP_005265798.1 | ||||
XM_005265742.2 | Intron | XP_005265799.1 | ||||
XM_017008178.1 | Intron | XP_016863667.1 | ||||
XM_017008179.1 | Intron | XP_016863668.1 | ||||
XM_017008180.1 | Intron | XP_016863669.1 |
Set Membership: |
HapMap |